6-Methyl-2-pyridinemethanol ≥98.0%
Supplier: TCI America
M1282-25G
M1282-5G
TCM1282-25GEA
880.06
CAD
TCM1282-25G
TCM1282-005G
6-Methyl-2-pyridinemethanol ≥98.0%
6-Methyl-2-pyridinemethanol
CAS Number: 1122-71-0
MDL Number: MFCD00023520
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 108
Freezing point (°C): 28
Other Product Names / Synonyms: 2-Hydroxymethyl-6-methylpyridine, 6-Hydroxymethyl-2-picoline
Formula:
C₇H₉NO Flash Pt: 108 °C |
MDL Number:
MFCD00023520 CAS Number: 1122-71-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...