(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt ≥97.0%

Supplier: TCI America

Synonyms: Methyl N-(Triethylammoniosulfonyl)carbamate, Methyl N-(Triethylammoniosulphonyl)carbamate, Burgess Reagent

M1279-25G M1279-1G M1279-5G
TCM1279-25GEA 823.18 CAD
TCM1279-25G TCM1279-001G TCM1279-5G
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt ≥97.0%
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt

CAS Number: 29684-56-8
MDL Number: MFCD00077815
Molecular Formula: C8H18N2O4S
Molecular Weight: 238.30
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 79
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Burgess Reagent, Methyl N-(Triethylammoniosulfonyl)carbamate

Formula: C₈H₁₈N₂O₄S
Melting Pt: 79 °C
MDL Number: MFCD00077815
CAS Number: 29684-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65bc5d0f2e8f4bd0c4a7c6b2f8ac1db0d33a809a&vid=65bc5d0f2e8f4bd0c4a7c6b2f8ac1db0d33a809a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR