4-Methoxybenzhydrol ≥98.0%

Supplier: TCI America

M1278-5G M1278-25G
TCM1278-005GEA 79 CAD
TCM1278-005G TCM1278-25G
4-Methoxybenzhydrol ≥98.0%
4-Methoxybenzhydrol

CAS Number: 720-44-5
MDL Number: MFCD00014398
Molecular Formula: C14H14O2
Molecular Weight: 214.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 65


Other Product Names / Synonyms: (4-Methoxyphenyl)(phenyl)methanol

Formula: C₁₄H₁₄O₂
Melting Pt: 65 °C
MDL Number: MFCD00014398
CAS Number: 720-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=922da9d393bd0073b18ca1ec5d2bb9c3263103c5&vid=922da9d393bd0073b18ca1ec5d2bb9c3263103c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR