5-Methoxyindole-2-carboxylic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

M1271-5G M1271-25G
TCM1271-005GEA 110.6 CAD
TCM1271-005G TCM1271-025G
5-Methoxyindole-2-carboxylic acid ≥97.0% (by HPLC, titration analysis)
5-Methoxyindole-2-carboxylic acid

CAS Number: 4382-54-1
MDL Number: MFCD00005614
Molecular Formula: C10H9NO3
Molecular Weight: 191.19
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal

Formula: C₁₀H₉NO₃
MDL Number: MFCD00005614
CAS Number: 4382-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5d11b1edc934824e92cc7b48fd0e26434d0d478&vid=e5d11b1edc934824e92cc7b48fd0e26434d0d478&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR