5-Methoxy-1H-indole-3-carbaldehyde

Supplier: TCI America

M1268-1G M1268-5G
TCM1268-001GEA 91.64 CAD
TCM1268-001G TCM1268-005G
5-Methoxy-1H-indole-3-carbaldehyde
5-Methoxy-1H-indole-3-carbaldehyde

CAS Number: 10601-19-1
MDL Number: MFCD00005623
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Form: Crystal
Color: Pale Yellow
Melting point (°C): 181


Other Product Names / Synonyms: 3-Formyl-5-methoxyindole

Formula: C10H9NO2
MW: 175.19 g/mol
Melting Pt: 181 °C
MDL Number: MFCD00005623
CAS Number: 10601-19-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b757b8f0b0451a0590c68fc49486be9a5c137bc&vid=4b757b8f0b0451a0590c68fc49486be9a5c137bc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR