Diethyl (methylthiomethyl)phosphonate ≥97.0%

Supplier: TCI America

Synonyms: Diethyl ((methylthio)methyl)phosphonate

M1208-5G M1208-25G
TCM1208-005GEA 81.05 CAD
TCM1208-005G TCM1208-25G
Diethyl (methylthiomethyl)phosphonate ≥97.0%
Diethyl (methylthiomethyl)phosphonate

CAS Number: 28460-01-7
MDL Number: MFCD00015131
Molecular Formula: C6H15O3PS
Molecular Weight: 198.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 101
Specific Gravity (20/20): 1.14


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (Methylthiomethyl)phosphonic Acid Diethyl Ester

Formula: C₆H₁₅O₃PS
MW: 198.220 g/mol
Boiling Pt: 120 °C
Density: 1.14 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00015131
CAS Number: 28460-01-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8c559288f9e01a224b02d2e94599d4cd3811736&vid=a8c559288f9e01a224b02d2e94599d4cd3811736&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR