4-(2-Methoxyethyl)phenol ≥98.0%

Supplier: TCI America

M1167-500G M1167-25G
TCM1167-500GEA 629.79 CAD
TCM1167-500G TCM1167-25G
4-(2-Methoxyethyl)phenol ≥98.0%
4-(2-Methoxyethyl)phenol

CAS Number: 56718-71-9
MDL Number: MFCD00017537
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 42

Formula: C₉H₁₂O₂
Boiling Pt: 125 °C
Melting Pt: 42 °C
MDL Number: MFCD00017537
CAS Number: 56718-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73e25b9ec159f3425706caba698f66b3cc49224e&vid=73e25b9ec159f3425706caba698f66b3cc49224e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR