3-Methyl-3-oxetanemethanol ≥96.0%
Supplier: TCI America
M1136-25G
M1136-5G
TCM1136-25GEA
101.12
CAD
TCM1136-25G
TCM1136-5G
3-Methyl-3-oxetanemethanol ≥96.0%
3-Methyl-3-oxetanemethanol
CAS Number: 3143-02-0
MDL Number: MFCD00010273
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 97
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: 2-(Hydroxymethyl)-2-methyl-1,3-epoxypropane, 3-(Hydroxymethyl)-3-methyloxetane
Formula:
C₅H₁₀O₂ Boiling Pt: 122 °C Density: 1.04 (20 °C) Flash Pt: 97 °C |
MDL Number:
MFCD00010273 CAS Number: 3143-02-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...