Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 4,6-O-Benzylidene-α-D-glucopyranoside, (4aR,6S,7R,8R,8aS)-6-Methoxy-2-phenylhexahydropyrano[3,2-d][1,3]dioxine-7,8-diol
M1125-25G
M1125-5G
TCM1125-025GEA
341.28
CAD
TCM1125-025G
TCM1125-005G
Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside ≥98.0%
Methyl-4,6-O-benzylidene-alpha-D-glucopyranoside
CAS Number: 3162-96-7
MDL Number: MFCD00006819
Molecular Formula: C14H18O6
Molecular Weight: 282.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 164
Specific rotation [a]20/D: 105 deg (C=2, CH3CN)
Formula:
C₁₄H₁₈O₆ MW: 282.29 g/mol Melting Pt: 164 °C |
MDL Number:
MFCD00006819 CAS Number: 3162-96-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...