1-Methyl-2-pyrrolecarboxaldehyde ≥97.0%
Supplier: TCI America
M1119-25G
M1119-5G
TCM1119-25GEA
88.48
CAD
TCM1119-25G
TCM1119-5G
1-Methyl-2-pyrrolecarboxaldehyde ≥97.0%
1-Methyl-2-pyrrolecarboxaldehyde
CAS Number: 1192-58-1
MDL Number: MFCD00003087
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 72
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: 2-Formyl-1-methylpyrrole, 1-Methylpyrrole-2-aldehyde
Formula:
C₆H₇NO Boiling Pt: 90 °C Density: 1.08 (20 °C) Flash Pt: 72 °C |
MDL Number:
MFCD00003087 CAS Number: 1192-58-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...