Methyl-β-D-galactopyranoside ≥98.0%
Supplier: TCI America
Synonyms:
Methyl-β-D-galactoside, (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol, (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
M1035-1G
M1035-5G
TCM1035-001GEA
77.42
CAD
TCM1035-001G
TCM1035-005G
Methyl-β-D-galactopyranoside ≥98.0%
Methyl-beta-D-galactopyranoside
CAS Number: 1824-94-8
MDL Number: MFCD00064357
Molecular Formula: C7H14O6
Molecular Weight: 194.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 178
Specific rotation [a]20/D: -16.5 deg (C=1.5, MeOH)
Formula:
C₇H₁₄O₆ MW: 194.18 g/mol Melting Pt: 178 °C |
MDL Number:
MFCD00064357 CAS Number: 1824-94-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...