3,4-(Methylenedioxy)phenylacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (1,3-Benzodioxol-5-yl)acetic acid

M1008-5G M1008-25G
TCM1008-005GEA 235.42 CAD
TCM1008-005G TCM1008-025G
3,4-(Methylenedioxy)phenylacetic acid ≥98.0% (by GC, titration analysis)
3,4-(Methylenedioxy)phenylacetic acid

CAS Number: 2861-28-1
MDL Number: MFCD00014576
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 129


Other Product Names / Synonyms: (1,3-Benzodioxol-5-yl)acetic Acid

Formula: C₉H₈O₄
MW: 180.16
Melting Pt: 129 °C
MDL Number: MFCD00014576
CAS Number: 2861-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a74130ba37379c6009fb32931f595d17f2b5299&vid=3a74130ba37379c6009fb32931f595d17f2b5299&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR