5-Methylpyrazine-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1003-5G M1003-25G
TCM1003-5GEA 30.02 CAD
TCM1003-5G TCM1003-025G
5-Methylpyrazine-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Methylpyrazine-2-carboxylic acid

CAS Number: 5521-55-1
MDL Number: MFCD00068241
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 166

Formula: C₆H₆N₂O₂
Melting Pt: 166 °C
MDL Number: MFCD00068241
CAS Number: 5521-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=268bf7bd8617bce029a11c76509ed84c7f31d9b5&vid=268bf7bd8617bce029a11c76509ed84c7f31d9b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR