4-(Methylthio)toluene ≥98.0%
Supplier: TCI America
M0949-10G
M0949-25G
TCM0949-010GEA
94.85
CAD
TCM0949-010G
TCM0949-25G
4-(Methylthio)toluene ≥98.0%
4-(Methylthio)toluene
CAS Number: 623-13-2
MDL Number: MFCD00008560
Molecular Formula: C8H10S
Molecular Weight: 138.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 85
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: 4-Methylthioanisole, Methyl p-Tolyl Sulfide
Formula:
C₈H₁₀S Boiling Pt: 94 °C Density: 1.03 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00008560 CAS Number: 623-13-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...