Methylthiomethyl p-Tolyl sulfone ≥98.0%

Supplier: TCI America

M0875-25G M0875-5G
TCM0875-025GEA 734.7 CAD
TCM0875-025G TCM0875-5G
Methylthiomethyl p-Tolyl sulfone ≥98.0%
Methylthiomethyl p-Tolyl sulfone

CAS Number: 59662-65-6
MDL Number: MFCD00010750
Molecular Formula: C9H12O2S2
Molecular Weight: 216.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 83


Product Categories: Synthetic Organic Chemistry, Sulfones
Other Product Names / Synonyms: Methylmercaptomethyl p-Tolyl Sulfone, Methyl p-Toluenesulfonylmethyl Sulfide, MT-sulfone

Formula: C₉H₁₂O₂S₂
Melting Pt: 83 °C
MDL Number: MFCD00010750
CAS Number: 59662-65-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9de9eab75fa0d4263121154bf39c57cd9c3a07e&vid=b9de9eab75fa0d4263121154bf39c57cd9c3a07e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR