6-Methylpicolinic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
M0873-25G
M0873-5G
TCM0873-25GEA
244.33
CAD
TCM0873-25G
TCM0873-5G
6-Methylpicolinic acid ≥98.0% (by HPLC, titration analysis)
6-Methylpicolinic acid
CAS Number: 934-60-1
MDL Number: MFCD00023481
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 130
Other Product Names / Synonyms: 6-Methylpicolinic Acid
Formula:
C₇H₇NO₂ Melting Pt: 130 °C |
MDL Number:
MFCD00023481 CAS Number: 934-60-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...