Diethyl mesoxalate (diethyl ketomalonate) ≥94.0%

Supplier: TCI America

Synonyms: Diethyl 2-oxomalonate

M0866-5G M0866-25G
TCM0866-005GEA 88.48 CAD
TCM0866-005G TCM0866-25G
Diethyl mesoxalate (diethyl ketomalonate) ≥94.0%
Diethyl mesoxalate (Diethyl ketomalonate)

CAS Number: 609-09-6
MDL Number: MFCD00009121
Molecular Formula: C7H10O5
Molecular Weight: 174.15
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Flash Point (°C): 110
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: Diethyl Ketomalonate, Mesoxalic Acid Diethyl Ester

Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 210 °C
Density: 1.14 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009121
CAS Number: 609-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8f0814e527d647112871cc3e01f84fcb7764c00&vid=b8f0814e527d647112871cc3e01f84fcb7764c00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR