Methyl-3-(methylthio)propionate
Supplier: TCI America
Synonyms:
Methyl3-(methylthio)propionate
M0862-25G
M0862-500G
TCM0862-025GEA
41.08
CAD
TCM0862-025G
TCM0862-500G
Methyl-3-(methylthio)propionate
Methyl 3-(methylthio)propionate
CAS Number: 13532-18-8
MDL Number: MFCD00008565
Molecular Formula: C5H10O2S
Molecular Weight: 134.19
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 81
Flash Point (°C): 72
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 3-(Methylthio)propionic Acid Methyl Ester
Formula:
C₅H₁₀O₂S Boiling Pt: 81 °C Density: 1.07 (20 °C) Flash Pt: 72 °C |
MDL Number:
MFCD00008565 CAS Number: 13532-18-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...