5-Nitro-1,3-benzodioxole ≥98.0%

Supplier: TCI America

M0848-500G M0848-25G
TCM0848-500GEA 1760.12 CAD
TCM0848-500G TCM0848-025G
5-Nitro-1,3-benzodioxole ≥98.0%
5-Nitro-1,3-benzodioxole

CAS Number: 2620-44-2
MDL Number: MFCD00005824
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 148
Flash Point (°C): 143


Other Product Names / Synonyms: 5-Nitro-1,3-benzodioxole

Formula: C₇H₅NO₄
Melting Pt: 148 °C
Flash Pt: 143 °C
MDL Number: MFCD00005824
CAS Number: 2620-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4154330782d6672d1adc4bf60b15e96ad001c2b&vid=d4154330782d6672d1adc4bf60b15e96ad001c2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR