2-Methyl-3-propylpyrazine ≥95.0%

Supplier: TCI America

M0841-5ML M0841-25ML
TCM0841-5MLEA 83.74 CAD
TCM0841-5ML TCM0841-25ML
2-Methyl-3-propylpyrazine ≥95.0%
2-Methyl-3-propylpyrazine

CAS Number: 15986-80-8
MDL Number: MFCD00010096
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 69
Specific Gravity (20/20): 0.97

Formula: C₈H₁₂N₂
Boiling Pt: 190 °C
Density: 0.97 (20 °C)
Flash Pt: 69 °C
MDL Number: MFCD00010096
CAS Number: 15986-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3cd8ac9b84452780eba3127e73283d911df860a&vid=d3cd8ac9b84452780eba3127e73283d911df860a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR