Maltitol ≥98.0%

Supplier: TCI America

Synonyms: (2S,3R,4R,5R)-4-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol

M0797-500G M0797-25G M0797-100G
TCM0797-500GEA 632 CAD
TCM0797-500G TCM0797-025G TCM0797-100G
Maltitol ≥98.0%
Maltitol

CAS Number: 585-88-6
MDL Number: MFCD00006600
Molecular Formula: C12H24O11
Molecular Weight: 344.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 151
Specific rotation [a]20/D: 107 deg (C=10, H2O)

Formula: C₁₂H₂₄O₁₁
Melting Pt: 151 °C
MDL Number: MFCD00006600
CAS Number: 585-88-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d59488ccdb416558e4c55d41e46ea89150afc4e7&vid=d59488ccdb416558e4c55d41e46ea89150afc4e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR