Tetrahydrofurfuryl methacrylate ≥98.0% stabilised
Supplier: TCI America
Synonyms:
(Tetrahydrofuran-2-yl)methyl methacrylate, Methacrylic Acid Tetrahydrofurfuryl Ester
(stabilized with MEHQ)
CAS Number: 2455-24-5
MDL Number: MFCD00014515
Molecular Formula: C9H14O3
Molecular Weight: 170.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 96
Specific Gravity (20/20): 1.04
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid Tetrahydrofurfuryl Ester
Formula:
C₉H₁₄O₃ Boiling Pt: 85 °C Density: 1.04 (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00014515 CAS Number: 2455-24-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...