3-Methyl-1,5-pentanediol ≥97.0%
Supplier: TCI America
M0710-25ML
M0710-500ML
TCM0710-025MLEA
55.3
CAD
TCM0710-025ML
TCM0710-500ML
3-Methyl-1,5-pentanediol ≥97.0%
3-Methyl-1,5-pentanediol
CAS Number: 4457-71-0
MDL Number: MFCD00039547
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 146
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: 1,5-Dihydroxy-3-methylpentane
Formula:
C₆H₁₄O₂ Boiling Pt: 150 °C Density: 0.97 (20 °C) Flash Pt: 146 °C |
MDL Number:
MFCD00039547 CAS Number: 4457-71-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...