N-Methyl-ε-caprolactam ≥97.0%

Supplier: TCI America

Synonyms: N-Methyl-2-azacycloheptanone, 1-Methylazepan-2-one

M0686-25G M0686-500G
TCM0686-025GEA 74.26 CAD
TCM0686-025G TCM0686-500G
N-Methyl-ε-caprolactam ≥97.0%
N-Methyl-epsilon-caprolactam

CAS Number: 2556-73-2
MDL Number: MFCD00003263
Molecular Formula: C7H13NO
Molecular Weight: 127.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 103
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: N-Methyl-2-azacycloheptanone

Formula: C₇H₁₃NO
Boiling Pt: 120 °C
Density: 1.02 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00003263
CAS Number: 2556-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f41ab3becd3b4ede7fef42fece32a18e8cbcf3d&vid=4f41ab3becd3b4ede7fef42fece32a18e8cbcf3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR