Maltitol ≥95.0%
Supplier: TCI America
Synonyms:
(2S,3R,4R,5R)-4-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol
M0601-25G
M0601-500G
TCM0601-025GEA
58.46
CAD
TCM0601-025G
TCM0601-500G
Maltitol ≥95.0%
Maltitol
CAS Number: 585-88-6
MDL Number: MFCD00006600
Molecular Formula: C12H24O11
Molecular Weight: 344.31
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 146
Specific rotation [a]20/D: 105 deg (C=10, H2O)
Formula:
C₁₂H₂₄O₁₁ Melting Pt: 146 °C |
MDL Number:
MFCD00006600 CAS Number: 585-88-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...