Cyclohexyl methacrylate ≥98.0% stabilized
Supplier: TCI America
M0594-500ML
M0594-25ML
TCM0594-500MLEA
159.09
CAD
TCM0594-500ML
TCM0594-025ML
Cyclohexyl methacrylate ≥98.0% stabilized
Cyclohexyl methacrylate
(stabilized with MEHQ)
CAS Number: 101-43-9
MDL Number: MFCD00014292
Molecular Formula: C10H16O2
Molecular Weight: 168.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 72
Flash Point (°C): 78
Specific Gravity (20/20): 0.97
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid Cyclohexyl Ester
Formula:
C₁₀H₁₆O₂ Boiling Pt: 72 °C Density: 0.97 (20 °C) Flash Pt: 78 °C |
MDL Number:
MFCD00014292 CAS Number: 101-43-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...