N-(4-Methoxybenzylidene)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M0582-25G
M0582-5G
TCM0582-025GEA
135.88
CAD
TCM0582-025G
TCM0582-5G
N-(4-Methoxybenzylidene)aniline ≥98.0% (by GC, titration analysis)
N-(4-Methoxybenzylidene)aniline
CAS Number: 836-41-9
MDL Number: MFCD00025836
Molecular Formula: C14H13NO
Molecular Weight: 211.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 63
Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: N-(p-Anisal)aniline, N-(p-Anisylidene)aniline
Formula:
C₁₄H₁₃NO Melting Pt: 63 °C |
MDL Number:
MFCD00025836 CAS Number: 836-41-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...