Methoxyhydroquinone ≥98.0%
Supplier: TCI America
M0539-25G
M0539-5G
TCM0539-25GEA
178.54
CAD
TCM0539-25G
TCM0539-5G
Methoxyhydroquinone ≥98.0%
Methoxyhydroquinone
CAS Number: 824-46-4
MDL Number: MFCD00013971
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 89
Other Product Names / Synonyms: 1,4-Dihydroxy-2-methoxybenzene
Formula:
C₇H₈O₃ Boiling Pt: 180 °C Melting Pt: 89 °C |
MDL Number:
MFCD00013971 CAS Number: 824-46-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...