Hexyl methacrylate ≥98.0% stabilized

Supplier: TCI America

M0503-25ML M0503-500ML
TCM0503-025MLEA 69.19 CAD
TCM0503-025ML TCM0503-500ML
Hexyl methacrylate ≥98.0% stabilized
Hexyl methacrylate

(stabilized with MEHQ)
CAS Number: 142-09-6
MDL Number: MFCD00015283
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 80
Specific Gravity (20/20): 0.89


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid Hexyl Ester

Formula: C₁₀H₁₈O₂
Boiling Pt: 88 °C
Density: 0.89 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00015283
CAS Number: 142-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ac6d78ef4f2ba62b491341da24c8c30f3727e68&vid=2ac6d78ef4f2ba62b491341da24c8c30f3727e68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR