3-Methyl-1H-pyrazol-5(4H)-one ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M0491-25G M0491-500G
TCM0491-025GEA 63.44 CAD
TCM0491-025G TCM0491-500G
3-Methyl-1H-pyrazol-5(4H)-one ≥98.0% (by titrimetric analysis)
3-Methyl-1H-pyrazol-5(4H)-one

CAS Number: 108-26-9
MDL Number: MFCD00020699
Molecular Formula: C4H6N2O
Molecular Weight: 98.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 222

Formula: C4H6N2O
Melting Pt: 222 °C
MDL Number: MFCD00020699
CAS Number: 108-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79c2a7cd9e87e3ad93e93e7a70ec3661f3db6c1a&vid=79c2a7cd9e87e3ad93e93e7a70ec3661f3db6c1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR