Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥95.0%

Supplier: TCI America

Synonyms: Molybdenum(VI)dioxy Acetylacetonate, Acetylacetone Molybdenum(VI)dioxy Salt, Molybdenum(VI) oxide bis(acetylacetonate), Molybdenum(VI) dioxide bis(acetylacetonate), Molybdenyl acetylacetonate, Bis(acetylacetonato)dioxomolybdenum(VI)

M0464-25G M0464-5G
TCM0464-025GEA 370.19 CAD
TCM0464-025G TCM0464-5G
Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥95.0%
Bis(2,4-pentanedionato)molybdenum(VI) dioxide

CAS Number: 17524-05-9
MDL Number: MFCD00011506
Molecular Formula: C10H14MoO6
Molecular Weight: 326.18
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: Acetylacetone Molybdenum(VI)dioxy Salt, Bis(acetylacetonato)dioxomolybdenum(VI), Molybdenum(VI)dioxy Acetylacetonate

Formula: C₁₀H₁₄MoO₆
MW: 326.18 g/mol
MDL Number: MFCD00011506
CAS Number: 17524-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e598d64b7ef8acadb7d67ae31c3cdeb254f03930&vid=e598d64b7ef8acadb7d67ae31c3cdeb254f03930&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR