4-(Methylthio)phenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M0441-250G
M0441-25G
TCM0441-250GEA
1319.3
CAD
TCM0441-250G
TCM0441-025G
4-(Methylthio)phenol ≥98.0% (by GC, titration analysis)
4-(Methylthio)phenol
CAS Number: 1073-72-9
MDL Number: MFCD00002351
Molecular Formula: C7H8OS
Molecular Weight: 140.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 86
Formula:
C₇H₈OS Boiling Pt: 154 °C Melting Pt: 86 °C |
MDL Number:
MFCD00002351 CAS Number: 1073-72-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...