2-(Methylthio)benzimidazole ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

M0438-25G M0438-5G
TCM0438-25GEA 289.14 CAD
TCM0438-25G TCM0438-5G
2-(Methylthio)benzimidazole ≥99.0% (by HPLC, titration analysis)
2-(Methylthio)benzimidazole

CAS Number: 7152-24-1
MDL Number: MFCD00005594
Molecular Formula: C8H8N2S
Molecular Weight: 164.23
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 202


Other Product Names / Synonyms: 2-Methylmercaptobenzimidazole

Formula: C₈H₈N₂S
Melting Pt: 202 °C
MDL Number: MFCD00005594
CAS Number: 7152-24-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9723b571daeafe2a05fd66d8f88255f86c5f9445&vid=9723b571daeafe2a05fd66d8f88255f86c5f9445&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR