Methyl-ɑ-D-mannopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

M0368-25G M0368-250G
TCM0368-025GEA 104.19 CAD
TCM0368-025G TCM0368-250G
Methyl-ɑ-D-mannopyranoside ≥98.0% (by HPLC)
Methyl-alpha-D-mannopyranoside

CAS Number: 617-04-9
MDL Number: MFCD00063262
Molecular Formula: C7H14O6
Molecular Weight: 194.18
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 192
Specific rotation [a]20/D: 79.5 deg (C=4, H2O)


Product Categories: Bioscience, Mannose, Glycosides
Other Product Names / Synonyms: alpha-Methyl-D-mannoside, Methyl alpha-D-Mannoside

Formula: C₇H₁₄O₆
Melting Pt: 192 °C
MDL Number: MFCD00063262
CAS Number: 617-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01ef1a79923fef11049dfd1f1a2daabd840cd62e&vid=01ef1a79923fef11049dfd1f1a2daabd840cd62e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR