Methyl cellulose, 20-30 mPa.s (20 °C) (C=2%, H₂O)
Supplier: TCI America
Synonyms:
Cellulose methyl ether
M0291-25G
M0291-500G
TCM0291-025GEA
51.29
CAD
TCM0291-025G
TCM0291-500G
Methyl cellulose, 20-30 mPa.s (20 °C) (C=2%, H₂O)
Methyl cellulose
CAS Number: 9004-67-5
MDL Number: MFCD00081763
Form: Crystal
Color: White
- Form: Crystal
- Color: White
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Cellulose, Natural Polymers (for DDS)
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...