Benzyl methacrylate ≥98.0% stabilised

Supplier: TCI America

Synonyms: Methacrylic Acid Benzyl Ester

M0279-500ML M0279-25ML
TCM0279-500MLEA 115.69 CAD
TCM0279-500ML TCM0279-025ML
Benzyl methacrylate ≥98.0% stabilised
Benzyl methacrylate

(stabilized with MEHQ)
CAS Number: 2495-37-6
MDL Number: MFCD00053684
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 115
Specific Gravity (20/20): 1.04


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid Benzyl Ester

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 106 °C
Density: 1.04 (20 °C)
Flash Pt: 115 °C
MDL Number: MFCD00053684
CAS Number: 2495-37-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f8958793c6fb52a2e7b5c9e5e87d95e82c3f5ca&vid=2f8958793c6fb52a2e7b5c9e5e87d95e82c3f5ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR