Methyl-α-D-glucopyranoside ≥98.0%

Supplier: TCI America

M0228-25G M0228-500G M0228-100G
TCM0228-025GEA 35.5 CAD
TCM0228-025G TCM0228-500G TCM0228-100G
Methyl-α-D-glucopyranoside ≥98.0%
Methyl α-D-glucopyranoside

CAS Number: 97-30-3
MDL Number: MFCD00064086
Molecular Formula: C7H14O6
Molecular Weight: 194.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 170
Specific rotation [a]20/D: 157.5 deg (C=10, H2O)


Product Categories: Bioscience, Glucose, Glycosides
Other Product Names / Synonyms: Methyl alpha-D-Glucoside

Formula: C₇H₁₄O₆
Melting Pt: 170 °C
MDL Number: MFCD00064086
CAS Number: 97-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a2c5b0219489badba2b8727c5dc13c69dea5d4d&vid=0a2c5b0219489badba2b8727c5dc13c69dea5d4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR