Methylenebis(thioglycolic acid) ≥96.0% (by titrimetric analysis)

Supplier: TCI America

M0218-5G M0218-25G
TCM0218-5GEA 132.72 CAD
TCM0218-5G TCM0218-025G
Methylenebis(thioglycolic acid) ≥96.0% (by titrimetric analysis)
Methylenebis(thioglycolic acid)

CAS Number: 2068-24-8
MDL Number: MFCD00135949
Molecular Formula: C5H8O4S2
Molecular Weight: 196.24
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 129

Formula: C₅H₈O₄S₂
Melting Pt: 129 °C
MDL Number: MFCD00135949
CAS Number: 2068-24-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c988bf86bad5c7924fc27a9292c62166b21af07f&vid=c988bf86bad5c7924fc27a9292c62166b21af07f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR