4,4'-Methylenebis(2,6-di-tert-butylphenol) ≥98.0%
Supplier: TCI America
Synonyms:
Bis(3,5-di-tert-butyl-4-hydroxyphenyl)methane
M0214-500G
M0214-25G
TCM0214-500GEA
219.78
CAD
TCM0214-500G
TCM0214-025G
4,4'-Methylenebis(2,6-di-tert-butylphenol) ≥98.0%
4,4'-Methylenebis(2,6-di-tert-butylphenol)
CAS Number: 118-82-1
MDL Number: MFCD00008822
Molecular Formula: C29H44O2
Molecular Weight: 424.67
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 155
Product Categories: Materials Chemistry, Monomers, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(3,5-di-tert-butyl-4-hydroxyphenyl)methane
Formula:
C₂₉H₄₄O₂ MW: 424.67 g/mol Melting Pt: 155 °C |
MDL Number:
MFCD00008822 CAS Number: 118-82-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...