(Methyleneamino)acetonitrile

Supplier: TCI America

Synonyms: alpha-Hydroformamine Cyanide, Methyleneglycinonitrile

M0212-25G M0212-500G
TCM0212-025GEA 47.4 CAD
TCM0212-025G TCM0212-500G
(Methyleneamino)acetonitrile
(Methyleneamino)acetonitrile

CAS Number: 109-82-0
MDL Number: MFCD00001889
Molecular Formula: C3H4N2
Molecular Weight: 68.08
Form: Crystal
Color: White
Melting point (°C): 129


Other Product Names / Synonyms: alpha-Hydroformamine Cyanide, Methyleneglycinonitrile

Formula: C₃H₄N₂
Melting Pt: 129 °C
MDL Number: MFCD00001889
CAS Number: 109-82-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=074e91cabbe7b5652b21a5c347287aa1c9829664&vid=074e91cabbe7b5652b21a5c347287aa1c9829664&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR