DL-Methionine sulfone 99,0

Supplier: TCI America

M0100-1G M0100-5G
TCM0100-001GEA 69.52 CAD
TCM0100-001G TCM0100-005G
DL-Methionine sulfone 99,0
DL-Methionine sulphone

CAS Number: 820-10-0
MDL Number: MFCD00025079
Molecular Formula: C5H11NO4S
Molecular Weight: 181.21
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 250


Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: 2-Amino-4-(methylsulfonyl)butyric Acid

Formula: C₅H₁₁NO₄S
MW: 181.21 g/mol
Melting Pt: 250 °C
MDL Number: MFCD00025079
CAS Number: 820-10-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73febbdb144c2198e00f8cde83c5fba4f872089f&vid=73febbdb144c2198e00f8cde83c5fba4f872089f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR