Poly(methyl methacrylate)

Supplier: TCI America

Synonyms: Methacrylic Acid Methyl Ester Polymer, Methyl Methacrylate Polymer, PMMA

M0088-25G M0088-500G
TCM0088-025GEA 44.98 CAD
TCM0088-025G TCM0088-500G
Poly(methyl methacrylate)
Poly(methyl methacrylate)

CAS Number: 9011-14-7
MDL Number: MFCD00134349
Molecular Formula: C5H8O2
Form: Crystal
Color: White


Other Product Names / Synonyms: Methacrylic Acid Methyl Ester Polymer, Poly(methyl Methacrylate), PMMA

Formula: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
MDL Number: MFCD00134349
CAS Number: 9011-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff06b8f87746788834baa963ce560946cbde0978&vid=ff06b8f87746788834baa963ce560946cbde0978&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR