Benzyl-(S)-(-)-lactate ≥98.0%
Supplier: TCI America
Synonyms:
Benzyl L-lactate, (S)-(-)-Lactic Acid Benzyl Ester, Benzyl (S)-Lactate
CAS Number: 56777-24-3
MDL Number: MFCD00209546
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Melting point (°C): 17
Flash Point (°C): 113
Specific Gravity (20/20): 1.12
Specific rotation [a]20/D: -19 deg (C=1, Acetone)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-Lactic Acid Benzyl Ester
Formula:
C₁₀H₁₂O₃ MW: 180.20 g/mol Boiling Pt: 141 °C Melting Pt: 17 °C Density: 1.12 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00209546 CAS Number: 56777-24-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...