D-tert-Leucine (H-D-Tle-OH) ≥98.0%

Supplier: TCI America

Synonyms: H-D-Tle-OH

L0157-5G L0157-1G
TCL0157-5GEA 147.1 CAD
TCL0157-5G TCL0157-001G
D-tert-Leucine (H-D-Tle-OH) ≥98.0%
D-tert-Leucine (H-D-Tle-OH)

CAS Number: 26782-71-8
MDL Number: MFCD00004265
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 9 deg (C=3, H2O)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-2-Amino-3,3-dimethylbutyric Acid, H-D-Tle-OH

Formula: C₆H₁₃NO₂
MW: 131.2 g/mol
MDL Number: MFCD00004265
CAS Number: 26782-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a49b3f75c1c66feb3f8dc1bbb485262cd866d922&vid=a49b3f75c1c66feb3f8dc1bbb485262cd866d922&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR