L(+)-Lyxose 98,0

Supplier: TCI America

L0153-1G L0153-5G
TCL0153-001GEA 180.12 CAD
TCL0153-001G TCL0153-005G
L(+)-Lyxose 98,0
L(+)-Lyxose

CAS Number: 1949-78-6
MDL Number: MFCD00064111
Molecular Formula: C5H10O5
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 14 deg (C=1, H2O)

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
MDL Number: MFCD00064111
CAS Number: 1949-78-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67f17f18ece0305c2a94e49792d2a5cf029b8a9f&vid=67f17f18ece0305c2a94e49792d2a5cf029b8a9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR