L-Leucinamide hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

L0094-5G L0094-1G
TCL0094-5GEA 142.2 CAD
TCL0094-5G TCL0094-001G
L-Leucinamide hydrochloride ≥98.0% (by titrimetric analysis)
L-Leucinamide hydrochloride

CAS Number: 10466-61-2
MDL Number: MFCD00013012
Molecular Formula: C6H14N2O
Molecular Weight: 166.65
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 256
Specific rotation [a]20/D: 9.8 deg (C=5, H2O)

Formula: C₆H₁₄N₂O
Melting Pt: 256 °C
MDL Number: MFCD00013012
CAS Number: 10466-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d8db04a58ca613f2ff043ef67b8617d66e52b3a&vid=9d8db04a58ca613f2ff043ef67b8617d66e52b3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR