Isobutyl laurate ≥96.0%

Supplier: TCI America

L0084-25ML L0084-500ML
TCL0084-025MLEA 50.56 CAD
TCL0084-025ML TCL0084-500ML
Isobutyl laurate ≥96.0%
Isobutyl laurate

CAS Number: 37811-72-6
MDL Number: MFCD00048424
Molecular Formula: C16H32O2
Molecular Weight: 256.43
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 150
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Lauric Acid Isobutyl Ester

Formula: C₁₆H₃₂O₂
Density: 0.86 (20 °C)
Flash Pt: 150 °C
MDL Number: MFCD00048424
CAS Number: 37811-72-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859c37955c870e06d5d046d0a02f61fd78a97688&vid=859c37955c870e06d5d046d0a02f61fd78a97688&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR