2,6-Lutidine-N-oxide ≥98.0%

Supplier: TCI America

Synonyms: 2,6-Dimethylpyridine N-Oxide

L0069-25G L0069-5G
TCL0069-025GEA 314.42 CAD
TCL0069-025G 76220-222
2,6-Lutidine-N-oxide ≥98.0%
2,6-Lutidine-N-oxide

CAS Number: 1073-23-0
MDL Number: MFCD00039715
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Specific Gravity (20/20): 1.10


Product Categories: Synthetic Organic Chemistry, Oxidation
Other Product Names / Synonyms: 2,6-Dimethylpyridine N-Oxide

Formula: C₇H₉NO
Boiling Pt: 145 °C
Density: 1.10 (20 °C)
MDL Number: MFCD00039715
CAS Number: 1073-23-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=748ab20847e1964ef36f13e0015c81f4a22f73a5&vid=748ab20847e1964ef36f13e0015c81f4a22f73a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR