(±)-α-Lipoic acid ≥99.0%

Supplier: TCI America

Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid

L0058-25G L0058-5G
TCL0058-025GEA 483.92 CAD
TCL0058-025G TCL0058-5G
(±)-α-Lipoic acid ≥99.0%
(±)-α-Lipoic acid

CAS Number: 1077-28-7
MDL Number: MFCD00005474
Molecular Formula: C8H14O2S2
Molecular Weight: 206.32
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 165
Storage Temperature: 0-10°C


Product Categories: Bioscience, Antioxidants, Crosslinkers
Other Product Names / Synonyms: DL-Thioctic Acid

Formula: C₈H₁₄O₂S₂
MW: 206.33 g/mol
Boiling Pt: 165 °C
MDL Number: MFCD00005474
CAS Number: 1077-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ddfa68dd0f122e094c321deae0ab60dc082b06f&vid=5ddfa68dd0f122e094c321deae0ab60dc082b06f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR