3,4-Dihydro-4-oxo-1,2,3-benzotriazine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

K0003-25G K0003-5G
TCK0003-025GEA 135.56 CAD
TCK0003-025G TCK0003-005G
3,4-Dihydro-4-oxo-1,2,3-benzotriazine ≥98.0% (by HPLC, titration analysis)
3,4-Dihydro-4-oxo-1,2,3-benzotriazine

CAS Number: 90-16-4
MDL Number: MFCD00052387
Molecular Formula: C7H5N3O
Molecular Weight: 147.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 218


Other Product Names / Synonyms: 1,2,3-Benzotriazine-4(3H)-one, 4-Ketobenzotriazine

Formula: C₇H₅N₃O
Melting Pt: 218 °C
MDL Number: MFCD00052387
CAS Number: 90-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe477c104919ed78d9ba583e04e4a0a2b8ec654a&vid=fe477c104919ed78d9ba583e04e4a0a2b8ec654a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR